Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MDH1 Antibody (CPTC-MDH1-1), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308427AF405
Description
MDH1 Monoclonal antibody specifically detects MDH1 in Human samples. It is validated for Western BlotSpecifications
MDH1 | |
Monoclonal | |
Alexa Fluor 405 | |
cytoplasmic, Cytosolic malate dehydrogenase, DEE88, Diiodophenylpyruvate reductase, EC 1.1.1.37, EC 1.1.1.96, HEL-S-32, malate dehydrogenase 1, NAD (soluble), Malate dehydrogenase, cytoplasmic, MDH1, MDHA, MDH-s, MGC:1375, MOR2, soluble malate dehydrogenase | |
Recombinant human full-length MDH1 protein (Uniprot: P40925) | |
100 μg | |
Cancer, Immunology, Stem Cell Markers, Virology Bacteria and Parasites | |
4190 | |
Store at 4°C in the dark. | |
IgG2a κ |
Western Blot | |
CPTC-MDH1-1 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction