Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MDH1 Antibody, DyLight 488, Novus Biologicals™

Sheep Polyclonal Antibody
Supplier: Novus Biologicals NBP177780G
Description
MDH1 Polyclonal antibody specifically detects MDH1 in Porcine samples. It is validated for Western Blot, ELISASpecifications
MDH1 | |
Polyclonal | |
Western Blot, ELISA | |
cytoplasmic, Cytosolic malate dehydrogenase, DEE88, Diiodophenylpyruvate reductase, EC 1.1.1.37, EC 1.1.1.96, HEL-S-32, malate dehydrogenase 1, NAD (soluble), Malate dehydrogenase, cytoplasmic, MDH1, MDHA, MDH-s, MGC:1375, MOR2, soluble malate dehydrogenase | |
MDH1 [Pig Heart] (Uniprot: P11708) | |
0.1 mL | |
Cancer, Immunology, Stem Cell Markers, Virology Bacteria and Parasites | |
4190 | |
Store at 4C in the dark. |
Western Blot, ELISA | |
DyLight 488 | |
50mM Sodium Borate | |
Sheep | |
Multi-step | |
RUO | |
Primary | |
Pig | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction