Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MDH1 Mouse anti-Human, Mouse, Clone: 1F9A2, Proteintech
SDP

Catalog No. 501736927 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6927 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6927 Supplier Proteintech Group Inc Supplier No. 665051IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MDH1 is localized to the cytoplasm and may play pivotal roles in the malate-aspartate shuttle that operates in the metabolic coordination between cytosol and mitochondria.

Specifications

Antigen MDH1
Applications Western Blot
Classification Monoclonal
Clone 1F9A2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene MDH1
Gene Accession No. P14152, P40925
Gene Alias Cytosolic malate dehydrogenase, MDH s, MDH1, MDHA, MOR2
Gene Symbols Mdh1
Host Species Mouse
Immunogen MDH1 Fusion Protein Ag8744
Purification Method Protein G
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17449, 4190
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.