Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MDH1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597964
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597964 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597964 Supplier Invitrogen™ Supplier No. PA597964
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MDH1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

MDH1 is localized to the cytoplasm and may play pivotal roles in the malate-aspartate shuttle that operates in the metabolic coordination between cytosol and mitochondria.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MDH1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 3.36 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene MDH1
Gene Accession No. P14152, P40925
Gene Alias B230377B03Rik; Cytosolic malate dehydrogenase; D17921; Diiodophenylpyruvate reductase; epididymis secretory protein Li 32; HEL-S-32; malate dehydrogenase (EC 1.1.1.37); malate dehydrogenase 1; malate dehydrogenase 1, NAD (soluble); malate dehydrogenase, cytoplasmic; malate dehydrogenase, cytoplasmic; malate dehydrogenase, peroxisomal; malate dehydrogenase, soluble; Malate dehydrogenase-like enzyme; Mdh; Mdh1; MDHA; Mdhl; MDH-s; MDL1; MGC:1375; Mor2; Mor-2; soluble malate dehydrogenase
Gene Symbols MDH1
Host Species Rabbit
Immunogen Recombinant Human Malate dehydrogenase, cytoplasmic protein (1-334AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17449, 4190
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.