Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MDH1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA550446
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA550446 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA550446 Supplier Invitrogen™ Supplier No. PA550446
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total MDH1 protein.

MDH1 is localized to the cytoplasm and may play pivotal roles in the malate-aspartate shuttle that operates in the metabolic coordination between cytosol and mitochondria.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MDH1
Applications Western Blot
Classification Polyclonal
Concentration 2.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene MDH1
Gene Accession No. P14152, P40925
Gene Alias B230377B03Rik; Cytosolic malate dehydrogenase; D17921; Diiodophenylpyruvate reductase; epididymis secretory protein Li 32; HEL-S-32; malate dehydrogenase (EC 1.1.1.37); malate dehydrogenase 1; malate dehydrogenase 1, NAD (soluble); malate dehydrogenase, cytoplasmic; malate dehydrogenase, cytoplasmic; malate dehydrogenase, peroxisomal; malate dehydrogenase, soluble; Malate dehydrogenase-like enzyme; Mdh; Mdh1; MDHA; Mdhl; MDH-s; MDL1; MGC:1375; Mor2; Mor-2; soluble malate dehydrogenase
Gene Symbols MDH1
Host Species Rabbit
Immunogen Fusion protein corresponding to a region derived from internal residues of human malate dehydrogenase 1, NAD.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17449, 4190
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.