Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MDH2 Mouse anti-Human, Clone: 2F5AF8, Abnova™

Catalog No. 89035517 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-035-517 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-035-517 Supplier Abnova Corporation Supplier No. MAB14119

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against human MDH2.

  • High specificity
  • High reactivity

Specifications

Antigen MDH2
Applications ELISA, Immunocytochemistry, Immunoprecipitation
Classification Monoclonal
Clone 2F5AF8
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against human MDH2.
Dilution ELISA (4 ug/mL) Immunocytochemistry (4 ug/mL) Immunoprecipitation The optimal working dilution should be determined by the end user.
Formulation In HEPES (0.02% sodium azide).
Gene Accession No. P40926
Gene Alias M-MDH/MDH/MGC:3559/MOR1
Gene Symbols MDH2
Host Species Mouse
Immunogen Human MDH2.
Purification Method Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4191
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.