Learn More
MDK Mouse anti-Human, Clone: OTI6C8, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF807679

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Midkine is a retinoic acid-responsive, heparin-binding growth factor expressed in various cell types during embryogenesis. It promotes angiogenesis, cell growth, and cell migration. Midkine is also expressed in several carcinomas, suggesting that it may play a role in tumorigenesis, perhaps through its effects on angiogenesis.Specifications
MDK | |
Monoclonal | |
Unconjugated | |
MDK | |
ARAP, MK, NEGF2 | |
Mouse | |
Affinity Chromatography | |
RUO | |
4192 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI6C8 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P21741 | |
MDK | |
Human recombit protein fragment corresponding to amino acids 21-143 of human MDK produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.