Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MDL-1/CLEC5A Antibody (226402), PE/Cy7, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB1639PECY7
Description
MDL-1/CLEC5A Monoclonal antibody specifically detects MDL-1/CLEC5A in Mouse samples. It is validated for Flow CytometrySpecifications
MDL-1/CLEC5A | |
Monoclonal | |
PE-Cyanine7 | |
PBS | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Mouse | |
Purified |
Flow Cytometry | |
226402 | |
Flow Cytometry | |
CLECSF5carbohydrate-recognition domain) lectin, superfamilymember 5, C-type lectin domain family 5, member A, C-type lectin superfamily member 5, MDL-1MGC138304, Myeloid DAP12-associating lectin 1, myeloid DAP12-associating lectin-1 | |
BaF3 mouse pro-B cell line transfected with mouse MDL-1/CLEC5A | |
0.5 mL | |
Immunology | |
23601 | |
Store at 4C in the dark. Do not freeze. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction