Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MDL-1/CLEC5A Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP243607
Description
Specifically detects MDL-1/CLEC5A in Human samples, and it is validated for Western Blotting
MDL-1/CLEC5A Polyclonal specifically detects MDL-1/CLEC5A in Human, Mouse samples. It is validated for Western Blot.Specifications
MDL-1/CLEC5A | |
Polyclonal | |
Western Blot 1:500-1:3000 | |
CLECSF5carbohydrate-recognition domain) lectin, superfamilymember 5, C-type lectin domain family 5, member A, C-type lectin superfamily member 5, MDL-1MGC138304, Myeloid DAP12-associating lectin 1, myeloid DAP12-associating lectin-1 | |
Rabbit | |
22 kDa | |
0.1 mL | |
Immunology | |
23601 | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
PBS (pH 7.0) and 20% Glycerol with 0.01% Thimerosal | |
CLEC5A | |
Recombinant protein encompassing a sequence within the center region of human CLEC5A. The exact sequence is proprietary. | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction