Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ MDL 100907
GREENER_CHOICE

Catalog No. 417310
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
41-731-0 10 mg
41-735-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 41-731-0 Supplier Tocris Bioscience™ Supplier No. 4173/10
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective 5-HT2A antagonist

MDL 100907 is a potent and selective 5-HT2A receptor antagonist (Ki = 0.36 nM, IC50 = 3.3-5.1 nM). MDL 100907 exhibits > 80-fold selectivity for 5-HT2A receptors over other serotonergic receptor subtypes. MDL 100907 is neuroprotective; protects cultured dopaminergic neurons from MPP+ induced toxicity. Antipsychotic agent in vivo.

Chemical Identifiers

CAS 139290-65-6
Molecular Formula C22H28FNO3

Specifications

Quantity 10 mg
Formula Weight 373.46
Molecular Weight (g/mol) 373.47
Chemical Name or Material MDL 100907
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.