Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MDM1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559164
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559164 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559164 Supplier Invitrogen™ Supplier No. PA559164
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NNEGVTNHTP VNENVELEHS TKVLSENVDN GLDRLLRKKA GLTVVPSYNA LRNSEYQRQF VWKTSKETAP AFAANQVFHN KSQFVPPFKG NSVIHETEYK R Highest antigen sequence identity to the following orthologs: Mouse - 67%, Rat - 66%.

This gene encodes a nuclear protein similar to the mouse double minute 1 protein. The mouse gene is located in double minute (DM) chromatin particles, is amplified in the mouse transformed 3T3 cell line, and the encoded protein is able to bind to p53. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MDM1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Mdm1
Gene Accession No. Q8TC05
Gene Alias Arrd2; double minute 1; MDM1; Mdm-1; Mdm1 nuclear protein; Mdm1 nuclear protein homolog; Mdm1 nuclear protein homolog (mouse); mdm4 transformed 3T3 cell double minute 1 protein; Mdm4, transformed 3T3 cell double minute 1, p53 binding protein; Mouse double minute 1; nuclear protein double minute 1; nuclear protein MDM1; transformed mouse 3T3 cell double minute 1
Gene Symbols Mdm1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MDM1. Recombinant protein control fragment (Product #RP-97700).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56890
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.