Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MDM2/HDM2 Antibody (HDM2-323) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB6001312
Description
MDM2/HDM2 Monoclonal specifically detects MDM2/HDM2 in Human, Mouse samples. It is validated for Western Blot, ELISA.Specifications
MDM2/HDM2 | |
Monoclonal | |
2.0 mg/mL | |
Western Blot 1-2 ug/ml, ELISA 1:100-1:2000 | |
MDM2 | |
Synthetic peptide conjugated to KLH, corresponding to amino acids 389-402 of Human MDM2. | |
0.1 mL | |
Cancer, Cell Cycle and Replication, Hypoxia, Neuroscience, p53 Pathway, Tumor Suppressors | |
4193 | |
Human, Mouse | |
IgG2a |
Western Blot, ELISA | |
HDM2-323 | |
Unconjugated | |
ACTFS, HDM2, HDMX, MDM2 oncogene, E3 ubiquitin protein ligase | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
MDM2 (HDM2-323) | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction