Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MDM2/HDM2 Antibody (MDM2/8221), Novus Biologicals™

Mouse Monoclonal Antibody
$216.50 - $443.50
Specifications
Antigen | MDM2/HDM2 |
---|---|
Clone | MDM2/8221 |
Concentration | 0.2 mg/ml |
Dilution | Immunohistochemistry-Paraffin 1-2 μg/mL |
Applications | Immunohistochemistry (Paraffin) |
Description
MDM2/HDM2 Monoclonal antibody specifically detects MDM2/HDM2 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
MDM2/HDM2 | |
0.2 mg/ml | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Cancer, Cell Cycle and Replication, Cellular Markers, Hypoxia, Neuroscience, p53 Pathway, Phospho Specific, Tumor Suppressors | |
10mM PBS with 0.05% BSA | |
4193 | |
IgG2 κ | |
Protein A or G purified |
MDM2/8221 | |
Immunohistochemistry-Paraffin 1-2 μg/mL | |
Monoclonal | |
Purified | |
RUO | |
Human | |
ACTFS, HDM2, HDMX, MDM2 oncogene, E3 ubiquitin protein ligase | |
Recombinant human MDM2/HDM2 protein fragment (around aa 1-300) (exact sequence is proprietary) | |
Primary | |
Store at 4&drg;C. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title