Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MDM2/HDM2, p Ser185 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB1001683
Description
MDM2/HDM2 Polyclonal specifically detects MDM2/HDM2 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunoprecipitation.Specifications
MDM2/HDM2 (p Ser185) | |
Polyclonal | |
Unconjugated | |
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide | |
ACTFS, HDM2, HDMX, MDM2 oncogene, E3 ubiquitin protein ligase | |
Rabbit | |
54.56 kDa | |
0.1 mg | |
Cancer, Cell Cycle and Replication, Hypoxia, Neuroscience, p53 Pathway, Phospho Specific, Tumor Suppressors | |
4193 | |
Human, Mouse | |
IgG |
Western Blot, ELISA, Immunoprecipitation | |
1 mg/ml | |
Western Blot 1:500-1:2000, ELISA 1:3000-1:12000, Immunoprecipitation 1:100 | |
P23804 | |
MDM2 | |
This affinity purified MDM2/HDM2 [p Ser185] Antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to an internal region near aa 175-200 of mouse MDM2/HDM2/HDM2. (Uniprot: P23804) | |
Affinity Purified | |
RUO | |
Primary | |
This affinity-purified antibody is directed against the phosphorylated form of mouse MDM2/HDM2 protein at the pS185 residue. The resultant affinity purified antibody was then cross-adsorbed against the non-phosphorylated form of the immunizing peptide. Reactivity occurs against Mouse MDM2/HDM2 pS185 protein and the antibody is specific for the phosphorylated form of the protein. Reactivity with non-phosphorylated mouse MDM2/HDM2 is minimal by ELISA and western blot. A BLAST analysis was used to suggest minimal cross reactivity with MDM2/HDM2 homologues from other sources. | |
Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction