Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MDM2/HDM2, p Ser186 Antibody (2G2 2.2), Alexa Fluor™ 647, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250242AF647
Description
MDM2/HDM2 Monoclonal specifically detects MDM2/HDM2 in Human samples. It is validated for Western Blot.Specifications
MDM2/HDM2 | |
Monoclonal | |
Alexa Fluor 647 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
MDM2 | |
Peptide QRKRHKS(p)DSIS. S(p) represent a phosphorylated serine. This peptide represents the portion of MDM2/HDM2 around Serine 186 | |
0.1 mL | |
Cancer, Cell Cycle and Replication, Cellular Markers, Hypoxia, Neuroscience, p53 Pathway, Tumor Suppressors | |
4193 | |
Store at 4C in the dark. | |
IgG1 |
Western Blot | |
2G2 2.2 | |
Western Blot | |
ACTFS, HDM2, HDMX, MDM2 oncogene, E3 ubiquitin protein ligase | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction