Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MDM2/HDM2, p Ser186 Antibody (2G2 2.2) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$196.10
Specifications
Antigen | MDM2/HDM2 |
---|---|
Clone | 2G2 2.2 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:100 - 1:2000, Immunoprecipitation |
Applications | Western Blot, Immunoprecipitation |
Description
MDM2/HDM2 Monoclonal antibody specifically detects MDM2/HDM2 in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
MDM2/HDM2 | |
1 mg/mL | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Mouse | |
Cancer, Cell Cycle and Replication, Cellular Markers, Hypoxia, Neuroscience, p53 Pathway, Tumor Suppressors | |
PBS | |
4193 | |
IgG1 | |
Protein A purified |
2G2 2.2 | |
Western Blot 1:100 - 1:2000, Immunoprecipitation | |
Monoclonal | |
Purified | |
RUO | |
Human | |
ACTFS, HDM2, HDMX, MDM2 oncogene, E3 ubiquitin protein ligase | |
Peptide QRKRHKS(p)DSIS. S(p) represent a phosphorylated serine. This peptide represents the portion of MDM2/HDM2 around Serine 186 | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title