Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MDM2 Mouse anti-Human, Unconjugated, Clone: MDM2/2414, Abnova™

Catalog No. 89337779 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-337-779 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-337-779 Supplier Abnova Corporation Supplier No. MAB21288
Add to Cart
Add to Cart

Mouse Monoclonal Antibody.

MDM2 Monoclonal antibody specifically detects MDM2 in Human samples. It is validated for WB, IHC-P.

Specifications

Antigen MDM2
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone MDM2/2414
Conjugate Unconjugated
Description Mdm2 p53 binding protein homolog (mouse)
Gene MDM2
Gene Alias HDMX, MGC71221, hdm2
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 126-254 of human MDM2
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 4193
Target Species Human
Content And Storage Store at 2 to 8°C
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.