Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MDM2BP Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987232
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7232 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7232 Supplier Bioss Supplier No. BS6388R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MTBP inhibits cell migration in vitro and suppresses the invasive behavior of tumor cells. The protein may play a role in MDM2-dependent TP53/p53 homeostasis in unstressed cells. The protein inhibits autoubiquitination of MDM2, thereby enhancing MDM2 stability. This promotes MDM2-mediated ubiquitination of TP53/p53 and its subsequent degradation.

Specifications

Antigen MDM2BP
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Mtbp
Gene Accession No. Q96DY7
Gene Alias AI429604; hdm2; HDMX; hMTBP; MDM2 (mouse double minute 2)-binding protein, 104kD; MDM2 binding protein; Mdm2, transformed 3T3 cell double minute 2, p53 binding protein binding protein, 104kDa; Mdm2, transformed 3T3 cell double minute p53 binding protein; Mdm2-binding protein; MDM2BP; MGC5370; MGC71221; mMTBP; MTBP
Gene Symbols Mtbp
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human MTBP/MDM2BP.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27085
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.