Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    MDMX Antibody, Alexa Fluor™ 647, Novus Biologicals™
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297044AF647
Description
MDMX Polyclonal specifically detects MDMX in Human samples. It is validated for Western Blot, ELISA.Specifications
| MDMX | |
| Polyclonal | |
| Western Blot, ELISA | |
| DKFZp781B1423, double minute 4 homolog, Double minute 4 protein, double minute 4, human homolog of; p53-binding protein, HDMX, Mdm2-like p53-binding protein, Mdm4 p53 binding protein homolog (mouse), Mdm4, transformed 3T3 cell double minute 4, p53 binding protein, Mdm4, transformed 3T3 cell double minute 4, p53 binding protein (mouse), MDM4-related protein 1, MDMXMGC132766, mouse double minute 4, human homolog of; p53-binding protein, MRP1, p53-binding protein Mdm4, protein Mdm4, Protein Mdmx | |
| Produced in rabbits immunized with purified, recombinant MDMX (NP_002384.2; Met1-Asp134). | |
| 0.1 mL | |
| Apoptosis, DNA Repair, Phospho Specific, Signal Transduction | |
| 4194 | |
| Store at 4°C in the dark. | |
| IgG | 
| Western Blot, ELISA | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction