Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MDMX Antibody, DyLight 550, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB100556R
Description
MDMX Polyclonal antibody specifically detects MDMX in Human samples. It is validated for Western BlotSpecifications
MDMX | |
Polyclonal | |
Western Blot | |
DKFZp781B1423, double minute 4 homolog, Double minute 4 protein, double minute 4, human homolog of; p53-binding protein, HDMX, Mdm2-like p53-binding protein, Mdm4 p53 binding protein homolog (mouse), Mdm4, transformed 3T3 cell double minute 4, p53 binding protein, Mdm4, transformed 3T3 cell double minute 4, p53 binding protein (mouse), MDM4-related protein 1, MDMXMGC132766, mouse double minute 4, human homolog of; p53-binding protein, MRP1, p53-binding protein Mdm4, protein Mdm4, Protein Mdmx | |
The immunogen recognized by this antibody maps to a region between residues 125 and 175 of human ortholog of mouse double minute 4 using the numbering given in Swiss-Prot entry O15151 (GeneID 4194). | |
0.1 mL | |
Apoptosis, DNA Repair, Phospho Specific, Signal Transduction | |
4194 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
DyLight 550 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction