Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MDMX Antibody (OTI4G5), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP245779
Description
Ensure accurate, reproducible results in Western Blot, Immunohistochemistry, Immunofluorescence
MDMX Monoclonal specifically detects MDMX in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
MDMX | |
Monoclonal | |
0.6 mg/mL | |
Western Blot 1:2000, Immunohistochemistry 1:150, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:150 | |
DKFZp781B1423, double minute 4 homolog, Double minute 4 protein, double minute 4, human homolog of; p53-binding protein, HDMX, Mdm2-like p53-binding protein, Mdm4 p53 binding protein homolog (mouse), Mdm4, transformed 3T3 cell double minute 4, p53 binding protein, Mdm4, transformed 3T3 cell double minute 4, p53 binding protein (mouse), MDM4-related protein 1, MDMXMGC132766, mouse double minute 4, human homolog of; p53-binding protein, MRP1, p53-binding protein Mdm4, protein Mdm4, Protein Mdmx | |
Mouse | |
54.7 kDa | |
0.1 mL | |
Apoptosis, DNA Repair, Signal Transduction | |
4194 | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG2b |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
OTI4G5 | |
Unconjugated | |
PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
MDM4 | |
Full length human recombinant protein of human MDM4(NP_002384) produced in HEK293T cell. | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Monkey | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction