Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MDMX Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Rabbit Polyclonal Antibody

Supplier:  Proteintech Group Inc 179141AP150UL

Encompass_Preferred

 View more versions of this product

Catalog No. 50-173-1872


May include imposed supplier surcharges.
Only null left
Add to Cart

Description

Description

The MDM2 protein is the primary regulator of p53 protein stability. MDMX is an MDM2-related protein that inhibits MDM2-mediated degradation of p53 via distinct associations with MDM2. The gene that encodes MDMX (also designated MDM4) is a target for amplification in maligt gliomas. ARF interacts with MDMX to sequester MDMX within the nucleolus. This sequestration of MDMX by ARF results in an increase in p53 transactivation. In addition, expression of MDMX can reverse MDM2-targeted degradation of p53 while maintaining suppression of p53 transactivation. Like MDM2, MDMX also binds p73 and stabilizes the level of p73. Therefore, in striking contrast to p53, the half-life of p73 is increased by binding to MDM2.
Specifications

Specifications

MDMX
Polyclonal
Unconjugated
MDM4
Double minute 4 protein, HDMX, Mdm2 like p53 binding protein, MDM4, MDMX, MRP1, p53 binding protein Mdm4, Protein Mdm4, Protein Mdmx
Rabbit
Antigen Affinity Chromatography
RUO
17248, 304798, 4194
-20°C
Liquid
Immunoprecipitation, Western Blot
0.23 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O15151, O35618, Q5XIN1
Mdm4
MDMX Fusion Protein Ag12319
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.