Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MDR1/ABCB1 Antibody (MRK16), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NP22701A70 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP22701A70 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP22701A70 Supplier Novus Biologicals Supplier No. NBP252701AF750
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MDR1/ABCB1 Monoclonal specifically detects MDR1/ABCB1 in Human samples. It is validated for Flow Cytometry, Immunofluorescence.

Specifications

Antigen MDR1/ABCB1
Applications Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone MRK16
Conjugate Alexa Fluor 750
Dilution Flow Cytometry, Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias ABC20, ATP-binding cassette sub-family B member 1, ATP-binding cassette, sub-family B (MDR/TAP), member 1, CD243, CD243 antigen, CLCS, colchicin sensitivity, doxorubicin resistance, EC 3.6.3, EC 3.6.3.44, GP170, IBD13, MDR1MGC163296, multidrug resistance protein 1, P-glycoprotein 1, P-gp, PGY1P-GP
Gene Symbols ABCB1
Host Species Mouse
Immunogen Drug resistant K-562 cells.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 5243
Test Specificity Recognizes a discontinuous extracellular epitope on P-glycoprotein.
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.