Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MDR1/ABCB1 Antibody (MRK16) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$304.50
Specifications
Antigen | MDR1/ABCB1 |
---|---|
Clone | MRK16 |
Dilution | Flow Cytometry 1:10 - 1:1000, Immunoprecipitation 1:10 - 1:500, Immunofluorescence 1:10 - 1:500, CyTOF-ready |
Applications | Flow Cytometry, Immunoprecipitation, Immunofluorescence, CyTOF |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP25270125
![]() |
Novus Biologicals
NBP2527010.025MG |
0.025 mg |
Each of 1 for $304.50
|
|
|||||
Description
MDR1/ABCB1 Monoclonal specifically detects MDR1/ABCB1 in Human samples. It is validated for Flow Cytometry, Immunoprecipitation, Immunofluorescence, CyTOF-ready.Specifications
MDR1/ABCB1 | |
Flow Cytometry 1:10 - 1:1000, Immunoprecipitation 1:10 - 1:500, Immunofluorescence 1:10 - 1:500, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Human | |
ABC20, ATP-binding cassette sub-family B member 1, ATP-binding cassette, sub-family B (MDR/TAP), member 1, CD243, CD243 antigen, CLCS, colchicin sensitivity, doxorubicin resistance, EC 3.6.3, EC 3.6.3.44, GP170, IBD13, MDR1MGC163296, multidrug resistance protein 1, P-glycoprotein 1, P-gp, PGY1P-GP | |
ABCB1 | |
IgG1 | |
Protein A or G purified |
MRK16 | |
Flow Cytometry, Immunoprecipitation, Immunofluorescence, CyTOF | |
Unconjugated | |
Mouse | |
Cancer, Membrane Trafficking and Chaperones | |
PBS with 0.02% Proclin 300. with 0.02% Proclin 300 | |
5243 | |
Drug resistant K-562 cells. | |
Primary | |
Recognizes a discontinuous extracellular epitope on P-glycoprotein. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title