Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MDR1/ABCB1 Antibody (MRK16), Janelia Fluor™ 549 - Chimeric, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP252654JF549
Description
MDR1/ABCB1 Monoclonal specifically detects MDR1/ABCB1 in Human samples. It is validated for Flow Cytometry, Immunofluorescence.Specifications
MDR1/ABCB1 | |
Monoclonal | |
Janelia Fluor 549 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
ABCB1 | |
Drug resistant K-562 cells. | |
0.1 mL | |
Primary | |
Recognizes a discontinuous extracellular epitope on P-glycoprotein. | |
Store at 4°C in the dark. | |
IgG κ |
Flow Cytometry, Immunofluorescence | |
MRK16 | |
Flow Cytometry, Immunofluorescence | |
ABC20, ATP-binding cassette sub-family B member 1, ATP-binding cassette, sub-family B (MDR/TAP), member 1, CD243, CD243 antigen, CLCS, colchicin sensitivity, doxorubicin resistance, EC 3.6.3, EC 3.6.3.44, GP170, IBD13, MDR1MGC163296, multidrug resistance protein 1, P-glycoprotein 1, P-gp, PGY1P-GP | |
Rabbit | |
Protein A or G purified | |
Cancer, Membrane Trafficking and Chaperones | |
5243 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction