Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MDR1/ABCB1 Antibody (OTI1A7), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272607F
Description
MDR1/ABCB1 Monoclonal specifically detects MDR1/ABCB1 in Human, Mouse, Rat, Goat samples. It is validated for Immunohistochemistry.Specifications
MDR1/ABCB1 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
ABCB1 | |
Recombinant protein fragment corresponding to amino acids 995-1280 of human MDR1/ABCB1 (NP_000918) produced in SF9 cell. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat, Goat | |
IgG2a |
Immunohistochemistry | |
OTI1A7 | |
Immunohistochemistry | |
ABC20, ATP-binding cassette sub-family B member 1, ATP-binding cassette, sub-family B (MDR/TAP), member 1, CD243, CD243 antigen, CLCS, colchicin sensitivity, doxorubicin resistance, EC 3.6.3, EC 3.6.3.44, GP170, IBD13, MDR1MGC163296, multidrug resistance protein 1, P-glycoprotein 1, P-gp, PGY1P-GP | |
Mouse | |
Affinity Purified | |
Cancer, Membrane Trafficking and Chaperones | |
5243.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction