Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MDR3/ABCB4 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP30323420UL
Description
MDR3/ABCB4 Polyclonal specifically detects MDR3/ABCB4 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
MDR3/ABCB4 | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
5244 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunofluorescence | |
Unconjugated | |
ABC21, ATP-binding cassette sub-family B member 4, ATP-binding cassette, sub-family B (MDR/TAP), member 4, EC 3.6.3, EC 3.6.3.44, MDR2, MDR3MDR2/3, multidrug resistance protein 3, multiple drug resistance 3, P glycoprotein 3/multiple drug resistance 3, PFIC-3, P-glycoprotein 3, P-glycoprotein-3/multiple drug resistance-3, PGY3GBD1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-110 of human MDR3/ABCB4 (NP_061337.1). | |
20 μL | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction