Learn More
MDS028 Antibody (OTI3C4), DyLight 488, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP272614G
Catalog No. NBP272614G
Only null left
Add to Cart
Description
Description
MDS028 Monoclonal specifically detects MDS028 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
MDS028
Monoclonal
DyLight 488
50mM Sodium Borate with 0.05% Sodium Azide
ITFG2
Full length recombinant protein of human MDS028 (NP_060933) produced in HEK293T cells.
0.1 mL
55846.0
Store at 4°C in the dark.
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
OTI3C4
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
integrin alpha FG-GAP repeat containing 2, integrin-alpha FG-GAP repeat-containing protein 2, MDS028
Mouse
Affinity Purified
Primary
Human, Mouse, Rat
IgG1
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.