Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ME1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596265
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596265 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596265 Supplier Invitrogen™ Supplier No. PA596265
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: LO2, MCF7, Mouse heart, Mouse brain, Mouse liver, Mouse kidney; Cellular Location: Cytoplasm Immunogen sequence: MEPEAPRRRH THQRGYLLTR NPHLNKDLAF TLEERQQLNI HGLLPPSFNS QEIQVLRVVK NFEHLNSDFD RYLLLMDLQD RNEKLFYRVL TSDIEKFMPI VYTPTVGLAC QQYSLVFRKP RGLFITIHDR GHIASVLNAW PEDVIKAIVV TDGERILGLG DLGCNGMGIP VGKLALYTAC GGMNPQECLP VILDVGTENE ELLKDPLYIG LRQRRVRGSE YDDFLDEFME AVSSKYGMNC LIQFEDFANV NAFRLLNKYR NQYCTFNDDI QGTASVAVAG LLAALRITKN KLSDQTILFQ.

ME1 encodes a cytosolic, NADP-dependent enzyme that generates NADPH for fatty acid biosynthesis. The activity of this enzyme, the reversible oxidative decarboxylation of malate, links the glycolytic and citric acid cycles. The regulation of expression for this gene is complex. Increased expression can result from elevated levels of thyroid hormones or by higher proportions of carbohydrates in the diet.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ME1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 5.48 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Me1
Gene Accession No. P06801, P13697, P48163
Gene Alias cytosolic malic enzyme 1; D9Ertd267e; HUMNDME; malate dehydrogenase; malate dehydrogenase (oxaloacetate-decarboxylating) (NADP(+)); malate dehydrogenase (oxaloacetate-decarboxylating) (NADP+); malate dehydrogenase decarboxylase (NADP+); malic enzyme; malic enzyme 1; malic enzyme 1, NADP(+)-dependent, cytoplasmic (cytosolic, soluble); malic enzyme 1, NADP(+)-dependent, cytosolic; malic enzyme 1, soluble; Malic enzyme, cytoplasmic; malic enzyme, supernatant; Mdh-1; ME1; MES; Mod1; Mod-1; NADP-dependent malic enzyme; NADP-dependent malic enzyme-like protein; NADP-ME; pyruvic-malic carboxylase
Gene Symbols Me1
Host Species Rabbit
Immunogen Recombinant protein (or fragment)
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17436, 24552, 4199
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.