Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ME2 Monoclonal Antibody (970CT2.1.1.2)
GREENER_CHOICE

Catalog No. PIMA537745
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA537745 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA537745 Supplier Invitrogen™ Supplier No. MA537745
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ME2 Monoclonal Antibody for Western Blot, IHC (P)

ME2 encodes a mitochondrial NAD-dependent malic enzyme, a homotetrameric protein, that catalyzes the oxidative decarboxylation of malate to pyruvate. It had previously been weakly linked to a syndrome known as Friedreich ataxia that has since been shown to be the result of mutation in a completely different gene. Certain single-nucleotide polymorphism haplotypes of this gene have been shown to increase the risk for idiopathic generalized epilepsy. Alternatively spliced transcript variants encoding different isoforms found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ME2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 970CT2.1.1.2
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene Me2
Gene Accession No. P23368, Q99KE1
Gene Alias AW120568; D030040L20Rik; malate dehydrogenase; malate dehydrogenase (oxaloacetate-decarboxylating); malic enzyme 2; malic enzyme 2, NAD(+)-dependent, mitochondrial; Me2; NAD-dependent malic enzyme, mitochondrial; NAD-ME; ODS1; pyruvic-malic carboxylase
Gene Symbols Me2
Host Species Mouse
Immunogen KLH conjugated synthetic peptide between 522-556 amino acids from the C-terminal region of human ME2.
Purification Method Protein G
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 107029, 4200
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.