Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MEA1 Mouse anti-Human, Rat, Clone: 1H8C2, Proteintech
SDP

Catalog No. 501736895 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6895 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6895 Supplier Proteintech Group Inc Supplier No. 664721IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MEA1 gene ontology annotations related to this gene include spermatogenesis.

Specifications

Antigen MEA1
Applications Western Blot
Classification Monoclonal
Clone 1H8C2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene MEA1
Gene Accession No. Q16626, Q5FVH7
Gene Alias HYS, male enhanced antigen 1, MEA, MEA 1, MEA1
Gene Symbols MEA1
Host Species Mouse
Immunogen MEA1 Fusion Protein Ag0375
Purification Method Protein G
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4201, 685131
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.