Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MEA1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5140145
This item is not returnable.
View return policy
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: GSGPAGYSYQ PLNQDPEQEE VELAPVGDGD VVADIQDRIQ ALGLHLPDPP LESEDEDEEG.
MEA1 gene ontology annotations related to this gene include spermatogenesis.
Specifications
MEA1 | |
Polyclonal | |
Unconjugated | |
Mea1 | |
HYS; male enhanced antigen 1; Male-enhanced antigen (H-Y structural gene); male-enhanced antigen 1; male-enhanced antigen-1; MEA; MEA1; Mea-1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
4201 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q16626 | |
Mea1 | |
Recombinant protein corresponding to Human MEA1. Recombinant protein control fragment (Product #RP-109315). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction