Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MEAF6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5116390
Description
MEAF6 Polyclonal Antibody for Western Blot
The screening of cDNA expression libraries from human tumors with serum antibody (SEREX) has proven to be a powerful method for identifying the repertoire of tumor antigens recognized by the immune system of cancer patients, referred to as the cancer immunome. In this regard, cancer/testis (CT) antigens are of particular interest because of their immunogenicity and restricted expression patterns. Synovial sarcomas are striking with regard to CT antigen expression, however, highly expressed in sarcoma, CT antigens do not induce frequent humoral immune responses in sarcoma patients. Sera from two patients were used to immunoscreen cDNA libraries from two synovial sarcoma cell lines and normal testis, resulting in the identification of 113 distinct antigens.
Specifications
| MEAF6 | |
| Polyclonal | |
| Unconjugated | |
| MEAF6 | |
| 2310005N01Rik; 2810036M01Rik; C1orf149; C3H1orf149; CDABP0189; CENP-28; centromere protein 28; chromatin modification-related protein MEAF6; chromatin modification-related protein MEAF6-like; EAF6; Esa1-associated factor 6 homolog; Esa1p-associated factor 6 homolog; fj63g06; FLJ11730; meaf6; MYST/Esa1 associated factor 6; MYST/Esa1-associated factor 6; NY-SAR-91; Protein EAF6 homolog; RGD1310440; RP3-423B22.2; sarcoma antigen NY-SAR-91; wu:fj63g06; zgc:100869 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 64769 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q9HAF1 | |
| MEAF6 | |
| KLH conjugated synthetic peptide between 120-154 amino acids from human MEAF6. | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction