Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mebendazole 98.0+%, TCI America™
SDP

Catalog No. M227325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M227325G 25 g
M22735G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M227325G Supplier TCI America Supplier No. M227325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 31431-39-7
Molecular Formula C16H13N3O3
Molecular Weight (g/mol) 295.30
MDL Number MFCD00057872
InChI Key OPXLLQIJSORQAM-UHFFFAOYSA-N
Synonym mebendazole, vermox, telmin, mebenvet, pantelmin, vermirax, mebenoazole, ovitelmin, bantenol, mebutar
PubChem CID 4030
ChEBI CHEBI:6704
IUPAC Name methyl N-(6-benzoyl-1H-1,3-benzodiazol-2-yl)carbamate
SMILES COC(=O)NC1=NC2=CC=C(C=C2N1)C(=O)C1=CC=CC=C1

Specifications

Melting Point 287°C
Color White-Yellow
Quantity 25 g
Formula Weight 295.30
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Mebendazole
RTECSNumber EY8600000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.