Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Meclizine dihydrochloride
GREENER_CHOICESDP

Catalog No. 424550 Shop All Novus Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
42-455-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 42-455-0 Supplier R&D Systems™ Supplier No. 4245/50
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Human pregnane X receptor agonist; H1 antagonist

Meclizine dihydrochloride is a human pregnane X receptor (hPXR) agonist; it is a more potent activator of hPXR than rat PXR. Meclizine increases hPXR target gene expression in human hepatocyte primary cultures. It also acts as a histamine H1 receptor antagonist. Meclizine attenuates mitochondrial respiration and increases glycolysis. The product inhibits rotenone-induced cell death in stem cell-derived dopaminergic neuronal culture and confers neuroprotection in models of polyglutamine (polyQ) toxicity.

Chemical Identifiers

CAS 1104-22-9
Molecular Formula C25H29Cl3N2

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 463.87
Purity >99%
Molecular Weight (g/mol) 463.871
Chemical Name or Material Meclizine dihydrochloride
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.