Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Meclizine Hydrochloride, USP, 97-102%, Spectrum™ Chemical
SDP

Catalog No. 18604250
Click to view available options
Quantity:
100 g
Packaging:
Poly Pail

M3985, 1104-22-9, C25H27ClN2·2HCl

Spectrum™ Chemical Meclizine Hydrochloride, USP

Chemical Identifiers

CAS 1104-22-9
Molecular Formula C25H29Cl3N2
Molecular Weight (g/mol) 463.87
InChI Key VCTHNOIYJIXQLV-UHFFFAOYNA-N
IUPAC Name dihydrogen 1-[(4-chlorophenyl)(phenyl)methyl]-4-[(3-methylphenyl)methyl]piperazine dichloride
SMILES [H+].[H+].[Cl-].[Cl-].CC1=CC=CC(CN2CCN(CC2)C(C2=CC=CC=C2)C2=CC=C(Cl)C=C2)=C1

Specifications

CAS Max % 100%
Ignition Residue 0.1%
Packaging Poly Pail
Quantity 100 g
Percent Purity 97 to 102%
Grade USP
Physical Form Crystalline Powder or Lumps
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.