Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Meclofenamate Sodium, USP, 97-103%, Spectrum™ Chemical
SDP

Catalog No. 18604256 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
1 kg
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-604-256 1 kg Amber Glass Bottle
18-604-252 1 g Amber Glass Bottle
18-604-253 5 g Amber Glass Bottle
18-604-254 25 g Amber Glass Bottle
18-604-255 100 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-604-256 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. M11021KGBL

M1102, 6385-02-0, C14H10Cl2NNaO2·H2O

Spectrum™ Chemical Meclofenamate Sodium, USP is used as a drug in the treatment of dysmenorrhea, muscular pain, and arthritis. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 6385-02-0
Molecular Formula C14H10Cl2NNaO2
Molecular Weight (g/mol) 318.13
InChI Key OGPIIGMUPMPMNT-UHFFFAOYSA-M
IUPAC Name sodium 2-[(2,6-dichloro-3-methylphenyl)amino]benzoate
SMILES [Na+].CC1=CC=C(Cl)C(NC2=CC=CC=C2C([O-])=O)=C1Cl

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Quantity 1 kg
Percent Purity 97 to 103%
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.