Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MeCP2 Antibody (5H12), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP260787
Description
MeCP2 Monoclonal specifically detects MeCP2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
MeCP2 | |
Monoclonal | |
Unconjugated | |
PBS with 50% glycerol with 5mM Sodium Azide | |
MECP2 | |
Full length human recombinant MeCP2. Accession # P51608 | |
Protein G purified | |
Cancer, Chromatin Research, Neuroscience, Phospho Specific | |
4204 | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
5H12 | |
Western Blot 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500 | |
AUTSX3, DKFZp686A24160, MeCp-2 protein, mental retardation, X-linked 79, methyl CpG binding protein 2 (Rett syndrome), X-linked 16 | |
Mouse | |
75 kDa | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction