Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MeCP2 Monoclonal Antibody (4H7), Active Motif™

Rat Monoclonal Antibody
$159.90 - $651.90
Specifications
| Antigen | MeCP2 |
|---|---|
| Clone | 4H7 |
| Concentration | 1 mg/mL |
| Applications | ChIP Assay, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-3308
![]()
|
Active Motif
61292 |
10 μg |
Each for $159.90
|
|
|||||
50-199-3307
![]()
|
Active Motif
61291 |
100 μg |
Each for $651.90
|
|
|||||
Description
MECP2 belongs to a family of nuclear proteins (including MBD1, MBD2, MBD3 and MBD4) that have a methyl-CpG binding domain. MECP2 is capable of binding specifically to methylated DNA. DNA methylation is the major modification of eukaryotic genomes and plays an essential role in mammalian development. MECP2 can also repress transcription from methylated gene promoters. In contrast to other MBD family members, MECP2 is X-linked and subject to X inactivation. MECP2 is dispensible in stem cells, but is essential for embryonic development. MECP2 gene mutations are the cause of most cases of Rett syndrome, a progressive neurologic developmental disorder and one of the most common causes of mental retardation in females.Specifications
| MeCP2 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 30% glycerol and 0.035% sodium azide; pH 7.5 | |
| 1500041B07Rik; AUTSX3; D630021H01Rik; DKFZp686A24160; Mbd5; MECP2; MeCp-2 protein; methyl CpG binding protein 2; methyl-CpG binding protein 2; Methyl-CpG-binding protein 2; MRX16; MRX79; MRXS13; MRXSL; PPMX; RS; RTS; RTT; testis tissue sperm-binding protein Li 41a; WBP10 | |
| Mecp2 | |
| IgG2a | |
| Protein G | |
| Antibody |
| 4H7 | |
| ChIP Assay, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Rat | |
| Human, Mouse, Rat | |
| P51608, Q00566, Q9Z2D6 | |
| 17257, 29386, 4204 | |
| This MeCP2 antibody was raised against a full-length protein corresponding to rat MeCP2. | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Mecp2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title