Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MECP2 Mouse anti-Human, Clone:8H4A5B9, Abnova™

Catalog No. 89333530 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-333-530 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-333-530 Supplier Abnova Corporation Supplier No. MAB17294
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant human MECP2.

DNA methylation is the major modification of eukaryotic genomes and plays an essential role in mammalian development. Human proteins MECP2, MBD1, MBD2, MBD3, and MBD4 comprise a family of nuclear proteins related by the presence in each of a methyl-CpG binding domain (MBD). Each of these proteins, with the exception of MBD3, is capable of binding specifically to methylated DNA. MECP2, MBD1 and MBD2 can also repress transcription from methylated gene promoters. In contrast to other MBD family members, MECP2 is X-linked and subject to X inactivation. MECP2 is dispensible in stem cells, but is essential for embryonic development. MECP2 gene mutations are the cause of most cases of Rett syndrome, a progressive neurologic developmental disorder and one of the most common causes of mental retardation in females. [provided by RefSeq]

Specifications

Antigen MECP2
Applications ELISA, Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 8H4A5B9
Conjugate Unconjugated
Description methyl CpG binding protein 2 (Rett syndrome)
Dilution ELISA (1:10000)
Flow Cytometry (1:200-400)
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:200-1000)
Immunofluorescence (1:200-1000)
Western Blot (1:500-1:2000)
The optimal working dilution should be determined by th
Formulation In PBS (0.05% sodium azide)
Gene MECP2
Gene Alias AUTSX3, DKFZp686A24160, MRX16, MRX79, MRXS13, MRXSL, PPMX, RTS, RTT
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 7-148 of of human MECP2 from E. coli.
Quantity 100 μg
Gene ID (Entrez) 4204
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.