Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MECP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA1888 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA1888 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA1888 Supplier Invitrogen™ Supplier No. PA1888
  • $673.11
    $481.65 / Each of 1

    Save $191.46

    28% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

PA1-888 detects methyl CpG binding protein 2 (MeCP2) from human, mouse and rat cells and tissues. PA1-888 has been successfully used in Western blot procedures. By Western blot, this antibody detects an ~56 kDa protein representing MeCP2 from AtT20 cell extract. PA1-888 immunizing peptide corresponds to amino acid residues 469-486 from human MeCP2. This sequence is completely conserved in rat and mouse MeCP2. PA1-888 immunizing peptide (Cat. # PEP-121) is available for use in neutralization and control experiments.

MECP2 belongs to a family of nuclear proteins (including MBD1, MBD2, MBD3 and MBD4) that have a methyl-CpG binding domain. MECP2 is capable of binding specifically to methylated DNA. DNA methylation is the major modification of eukaryotic genomes and plays an essential role in mammalian development. MECP2 can also repress transcription from methylated gene promoters. In contrast to other MBD family members, MECP2 is X-linked and subject to X inactivation. MECP2 is dispensible in stem cells, but is essential for embryonic development. MECP2 gene mutations are the cause of most cases of Rett syndrome, a progressive neurologic developmental disorder and one of the most common causes of mental retardation in females.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MECP2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA and 0.05% sodium azide
Gene Mecp2
Gene Accession No. P51608, Q00566, Q9Z2D6
Gene Alias 1500041B07Rik; AUTSX3; D630021H01Rik; DKFZp686A24160; Mbd5; MECP2; MeCp-2 protein; methyl CpG binding protein 2; methyl-CpG binding protein 2; methyl-CpG-binding protein 2; MRX16; MRX79; MRXS13; MRXSL; PPMX; RS; RTS; RTT; testis tissue sperm-binding protein Li 41a; WBP10
Gene Symbols Mecp2
Host Species Rabbit
Immunogen Synthetic Peptide: C P(469) N R E E P V D S R T P V T E R V S(486).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17257, 29386, 4204
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.