Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MECP2 Polyclonal Antibody
MECP2 Polyclonal Antibody
Supplier: Thermo Scientific PA581220
Description
MECP2 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

Specifications
MECP2 | |
Polyclonal | |
Unconjugated | |
Mecp2 | |
1500041B07Rik; AUTSX3; D630021H01Rik; DKFZp686A24160; Mbd5; MECP2; MeCp-2 protein; methyl CpG binding protein 2; methyl-CpG binding protein 2; methyl-CpG-binding protein 2; MRX16; MRX79; MRXS13; MRXSL; PPMX; RS; RTS; RTT; testis tissue sperm-binding protein Li 41a; WBP10 | |
Rabbit | |
Antigen Affinity Chromatography, Protein A | |
RUO | |
4204 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with no preservative | |
P51608 | |
Mecp2 | |
A synthetic peptide corresponding to the C-terminus of the Human MECP2. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction