Learn More
MECP2 Rabbit anti-Human, CoraLite 555, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc CL55510861100UL
Description
The MECP2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human MECP2. This antibody recognizes human, mouse antigen. The MECP2 antibody has been validated for the following applications: FC (Intra), IF analysis. This antibody is coralite™555.
Specifications
| MECP2 | |
| Polyclonal | |
| Rabbit | |
| Primary | |
| Human, Mouse |
| Immunofluorescence, Flow Cytometry (Intracellular Staining) | |
| CoraLite 555 | |
| 100 μL | |
| 4204 | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.