Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ MeCP2, Rabbit, Unlabeled, Polyclonal,

Catalog No. ABN1728MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
ABN1728MI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABN1728MI Supplier MilliporeSigma™ Supplier No. ABN1728
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Methyl-CpG-binding protein 2 (UniProt P51608; also known as MeCp-2 protein, MeCp2) is encoded by the MECP2 (also known as AS, MRXS13, MRXSL, PPMX, RTT, AUTSX3) gene (Gene ID 4204) in human. Methyl-CpG-binding proteins that are involved in gene expression regulation by targeting methylated DNA residues in gene promoters via a conserved methyl-CpG-binding domain (MBD; a.a. 90-162 of human MeCp2). Upon binding methylated CpG sites on the DNA strands, transcriptional repression domain (TRD; a.a. 207-492) then mediates association with SIN3A to recruit histone deacetylases (HDAC). The TRD region also contains a nuclear localization signal (NLS) sequence (a.a. 255-271) and a nuclear receptor co-repressor 2/NCoR2-interacting site (a.a. 269-309). MeCp2 protein is found in all cells in the body, including the brain, where it is found in high concentrations in neurons and is associated with maturation of the central nervous system (CNS) and in forming synaptic contacts. In contrast to other MBD family members, MECP2 is X-linked and subject to X chromosome inactivation. MECP2 gene mutations are linked to Rett syndrome (RTT), a progressive neurologic developmental disorder and one of the most common causes of mental retardation in females.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MeCP2
Applications Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified rabbit polyclonal antibody in buffer containing PBS with 0.03% Sodium Azide.
Gene MECP2, AS, MRXS13, MRXSL, PPMX, RTT, AUTSX3
Gene Accession No. P51608
Host Species Rabbit
Immunogen Linear peptide corresponding to human MeCp2 C-terminal end sequence.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_004983
Test Specificity This polyclonal antibody targets MeCp2 C-terminal end sequence conserved between spliced isoform A (isoform Beta) and isoform B (isoform Alpha).
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.