Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MED10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111163
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111163 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111163 Supplier Invitrogen™ Supplier No. PA5111163
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FEYIDQGRNP QLYTKECLER ALAKNEQVKG KIDTMKKFKS LLIQELSKVF PEDMAKYRSI RGEDHPPS Highest antigen sequence indentity to the following orthologs: Mouse - 100%, Rat - 100%.

MED10 is a component of the Mediator complex, a coactivator involved in the regulated transcription of nearly all RNA polymerase II-dependent genes. Mediator functions as a bridge to convey information from gene-specific regulatory proteins to the basal RNA polymerase II transcription machinery. Mediator is recruited to promoters by direct interactions with regulatory proteins and serves as a scaffold for the assembly of a functional preinitiation complex with RNA polymerase II and the general transcription factors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MED10
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.20 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MED10
Gene Accession No. Q9BTT4
Gene Alias AA959813; AI385605; C78613; D13Wsu50e; L6; Med10; Mediator complex subunit 10; mediator of RNA polymerase II transcription subunit 10; mediator of RNA polymerase II transcription, subunit 10; mediator of RNA polymerase II transcription, subunit 10 homolog (NUT2, S. cerevisiae); NUT2; RGD1310128; Transformation-related gene 17 protein; transformation-related gene 20 protein; TRG17; TRG-17; TRG20; TRG-20
Gene Symbols MED10
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human MED10. Recombinant protein control fragment (Product #RP-100144).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84246
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.