Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MED11 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA575695
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
In mammalian cells, transcription is regulated in part by high molecular weight co-activating complexes that mediate signals between transcriptional activators and RNA polymerase II (Pol II). The mediator complex is one such multiprotein structure that functions as a bridge between regulatory proteins and Pol II, thereby regulating Pol II-dependent transcription. Med11 (mediator complex subunit 11) is a 117 amino acid nuclear protein and component of the mediator complex. The gene encoding Med11 maps to human chromosome 17, which comprises over 2.5% of the human genome and encodes over 1,200 genes. Two key tumor suppressor genes are associated with chromosome 17, namely, p53 and BRCA1. Malfunction or loss of p53 expression is associated with malignant cell growth and Li-Fraumeni syndrome. Like p53, BRCA1 is directly involved in DNA repair, and is associated with predisposition to cancers of the ovary, colon, prostate gland and fallopian tubes.
Specifications
MED11 | |
Polyclonal | |
Unconjugated | |
MED11 | |
1110030J09Rik; AI465144; AW545069; HSPC296; MED11; Mediator complex subunit 11; mediator of RNA polymerase II transcription subunit 11; mediator of RNA polymerase II transcription, subunit 11 homolog; RGD1563202 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
287456, 400569, 66172 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9D8C6, Q9P086 | |
MED11 | |
Synthetic peptide corresponding to residues in Human MED11. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction