Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MED11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103914
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103914 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103914 Supplier Invitrogen™ Supplier No. PA5103914
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total MED11.

In mammalian cells, transcription is regulated in part by high molecular weight co-activating complexes that mediate signals between transcriptional activators and RNA polymerase II (Pol II). The mediator complex is one such multiprotein structure that functions as a bridge between regulatory proteins and Pol II, thereby regulating Pol II-dependent transcription. Med11 (mediator complex subunit 11) is a 117 amino acid nuclear protein and component of the mediator complex. The gene encoding Med11 maps to human chromosome 17, which comprises over 2.5% of the human genome and encodes over 1,200 genes. Two key tumor suppressor genes are associated with chromosome 17, namely, p53 and BRCA1. Malfunction or loss of p53 expression is associated with malignant cell growth and Li-Fraumeni syndrome. Like p53, BRCA1 is directly involved in DNA repair, and is associated with predisposition to cancers of the ovary, colon, prostate gland and fallopian tubes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MED11
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene MED11
Gene Accession No. Q9D8C6, Q9P086
Gene Alias 1110030J09Rik; AI465144; AW545069; HSPC296; MED11; Mediator complex subunit 11; mediator of RNA polymerase II transcription subunit 11; mediator of RNA polymerase II transcription, subunit 11 homolog; RGD1563202
Gene Symbols MED11
Host Species Rabbit
Immunogen A synthesized peptide derived from human MED11(Accession Q9P086), corresponding to amino acid residues A13-L63.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 287456, 400569, 66172
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.