Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MED12 Antibody, DyLight 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB1002357UV
Description
MED12 Polyclonal specifically detects MED12 in Human, Mouse, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
MED12 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
ARC240, CAG repeat protein 45, CAGH45trinucleotide repeat containing 11 (THR-associated protein, 230kDa subunit), HOPAFGS1, KIAA0192FG syndrome 1, mediator complex subunit 12OPA-containing protein, OKSsubunit 12 homolog, thyroid hormone receptor-associated protein, 230 kDa subunit, Trap230, TRAP230mediator of RNA polymerase II transcription, subunit 12 homolog (S. cerevisiae), trinucleotide repeat containing 11 (THR-associated protein, 230 kDa subunit), Trinucleotide repeat-containing gene 11 protein | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse, Primate | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
DyLight 350 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
MED12 | |
The immunogen recognized by this antibody maps to a region between residue 2150 and the C-terminus (residue 2212) of human Mediator of RNA Polymerase II Transcription, subunit 12 (NP_005111.1). | |
100 μL | |
Signal Transduction, Vision | |
9968 | |
Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction