Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MED12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551852
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551852 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551852 Supplier Invitrogen™ Supplier No. PA551852
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LPTTMTGVMG LEPSSYKTSV YRQQQPAVPQ GQRLRQQLQA KIQSQGMLGQ SSVHQMTPSS SYGLQTSQGY TPYVSHVGLQ QHTGPAGTMV PPSYSSQPYQ STHPSTNPTL VDPTRHLQQR PSGYVHQQAP TYGHGLTS Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.

The initiation of transcription is controlled in part by a large protein assembly known as the preinitiation complex. A component of this preinitiation complex is a 1.2 MDa protein aggregate called Mediator. This Mediator component binds with a CDK8 subcomplex which contains the protein encoded by this gene, mediator complex subunit 12 (MED12), along with MED13, CDK8 kinase, and cyclin C.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MED12
Applications ChIP Assay, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Med12
Gene Accession No. Q93074
Gene Alias 230kDa; activator-recruited cofactor 240 kDa component; ARC240; CAG repeat protein 45; CAGH45; FGS1; HOPA; human opposite paired; Kiaa0192; Med12; MED12S; Mediator complex subunit 12; mediator of RNA polymerase II transcription subunit 12; mediator of RNA polymerase II transcription, subunit 12 homolog; Mopa; OHDOX; OKS; OPA1; OPA-1; OPA-containing protein; OPA-containing protein 1; putative mediator subunit 12; thyroid hormone receptor-associated protein complex 230 kDa component; thyroid hormone receptor-associated protein, 230 kDa subunit; TNRC11; TRAP230; trinucleotide repeat containing 11 (THR-associated protein); trinucleotide repeat containing 11 (THR-associated protein, 230 kDa subunit); trinucleotide repeat-containing gene 11 protein
Gene Symbols Med12
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MED12. Recombinant protein control fragment (Product #RP-92262).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9968
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.