Learn More
Invitrogen™ MED12 Recombinant Rabbit Monoclonal Antibody (BLR084G)
Description
Epitope retrieval with citrate buffer pH6.0 is recommended for FFPE cell sections.
Specifications
Specifications
| Antigen | MED12 |
| Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Clone | BLR084G |
| Concentration | 0.25 mg/mL |
| Conjugate | Unconjugated |
| Formulation | BBS with 0.1% BSA and 0.09% sodium azide; pH 8.2 |
| Gene | Med12 |
| Gene Accession No. | Q93074 |
| Gene Alias | 230kDa; activator-recruited cofactor 240 kDa component; ARC240; CAG repeat protein 45; CAGH45; FGS1; HOPA; human opposite paired; Kiaa0192; Med12; MED12S; Mediator complex subunit 12; mediator of RNA polymerase II transcription subunit 12; mediator of RNA polymerase II transcription, subunit 12 homolog; Mopa; OHDOX; OKS; OPA1; OPA-1; OPA-containing protein; OPA-containing protein 1; putative mediator subunit 12; thyroid hormone receptor-associated protein complex 230 kDa component; thyroid hormone receptor-associated protein, 230 kDa subunit; TNRC11; TRAP230; trinucleotide repeat containing 11 (THR-associated protein); trinucleotide repeat containing 11 (THR-associated protein, 230 kDa subunit); trinucleotide repeat-containing gene 11 protein |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.